MilliporeSigma
  • Regulation of Fas ligand expression and cell death by apoptosis-linked gene 4.
Regulation of Fas ligand expression and cell death by apoptosis-linked gene 4.
Nature medicine (1999-05-06)
E Lacana, L D'Adamio
ABSTRACT
Programmed cell death is a process required for the normal development of an organism. One of the best understood apoptotic pathways occurs in T lymphocytes and is mediated by Fas/Fas ligand (FasL) interaction. During studies of apoptosis induced by T cell-receptor engagement, we identified ALG-4F, a truncated transcript that prevents T cell-receptor-induced FasL upregulation and cell death. Overexpression of full-length ALG-4 induced transcription of FasL and, consequently, apoptosis. These results indicate that ALG-4 is necessary and sufficient for FasL expression. Fas/FasL interaction initiates cell death in many other systems, and its dysregulation is a mechanism by which several pathologic conditions arise. Understanding the molecular mechanisms of FasL regulation could be very useful in elucidating how these diseases develop and in identifying potential therapeutic targets.