MilliporeSigma
  • Oral administration of high molecular mass poly-gamma-glutamate induces NK cell-mediated antitumor immunity.
Oral administration of high molecular mass poly-gamma-glutamate induces NK cell-mediated antitumor immunity.
Journal of immunology (Baltimore, Md. : 1950) (2007-07-10)
Tae Woo Kim, Tae Young Lee, Hyun Cheol Bae, Jeong Ho Hahm, Yang Hyun Kim, Chung Park, Tae Heung Kang, Chul Joong Kim, Moon Hee Sung, Haryoung Poo
ABSTRACT
We analyzed the in vivo tumor regression activity of high molecular mass poly-gamma-glutamate (gamma-PGA) from Bacillus subtilis sups. chungkookjang. C57BL/6 mice were orally administered 10-, 100-, or 2000-kDa gamma-PGA or beta-glucan (positive control), and antitumor immunity was examined. Our results revealed higher levels of NK cell-mediated cytotoxicity and IFN-gamma secretion in mice treated with higher molecular mass gamma-PGA (2000 kDa) vs those treated with lower molecular mass gamma-PGA (10 or 100 kDa) or beta-glucan. We then examined the effect of oral administration of 10- or 2000-kDa gamma-PGA on protection against B16 tumor challenge in C57BL/6 mice. Mice receiving high molecular mass gamma-PGA (2000 kDa) showed significantly smaller tumor sizes following challenge with the MHC class I-down-regulated tumor cell lines, B16 and TC-1 P3 (A15), but not with TC-1 cells, which have normal MHC class I expression. Lastly, we found that gamma-PGA-induced antitumor effect was decreased by in vivo depletion of NK cells using mAb PK136 or anti-asialo GM1 Ab, and that was completely blocked in NK cell-deficient B6 beige mice or IFN-gamma knockout mice. Taken together, we demonstrated that oral administration of high molecular mass gamma-PGA (2000 kDa) generated significant NK cell-mediated antitumor activity in mice bearing MHC class I-deficient tumors.
Sigma-Aldrich
Poly-L-γ-glutamic acid sodium salt
Pricing and availability is not currently available.