MilliporeSigma
  • Enhancing electro-transformation competency of recalcitrant Bacillus amyloliquefaciens by combining cell-wall weakening and cell-membrane fluidity disturbing.
Enhancing electro-transformation competency of recalcitrant Bacillus amyloliquefaciens by combining cell-wall weakening and cell-membrane fluidity disturbing.
Analytical biochemistry (2010-10-19)
Guo-qiang Zhang, Peng Bao, Yun Zhang, Ai-hua Deng, Ning Chen, Ting-yi Wen
ABSTRACT
Bacillus amyloliquefaciens has been a major workhorse for the production of a variety of commercially important enzymes and metabolites for the past decades. Some subspecies of this bacterium are recalcitrant to exogenous DNA, and transformation with plasmid DNA is usually less efficient, thereby limiting the genetic manipulation of the recalcitrant species. In this work, a methodology based on electro-transformation has been developed, in which the cells were grown in a semicomplex hypertonic medium, cell walls were weakened by adding glycine (Gly) and DL-threonine (DL-Thr), and the cell-membrane fluidity was elevated by supplementing Tween 80. After optimization of the cell-loosening recipe by response surface methodology (RSM), the transformation efficiency reached 1.13 ± 0.34 × 10(7) cfu/μg syngeneic pUB110 DNA in a low conductivity electroporation buffer. Moreover, by temporary heat inactivation of the host restriction enzyme, a transformation efficiency of 8.94 ± 0.77 × 10(5) cfu/μg DNA was achieved with xenogeneic shuttle plasmids, a 10(3)-fold increase compared to that reported previously. The optimized protocol was also applicable to other recalcitrant B. amyloliquefaciens strains used in this study. This work could shed light on the functional genomics and subsequent strain improvement of the recalcitrant Bacillus, which are difficult to be transformed using conventional methods.
Sigma-Aldrich
DL-Threonine, ≥95% (titration)