MilliporeSigma
  • HU protein of Escherichia coli has a role in the repair of closely opposed lesions in DNA.
HU protein of Escherichia coli has a role in the repair of closely opposed lesions in DNA.
The Journal of biological chemistry (2003-05-16)
Mitsumasa Hashimoto, Barry Imhoff, Md Moshi Ali, Yoke W Kow
ABSTRACT
Closely opposed lesions form a unique class of DNA damage that is generated by ionizing radiation. Improper repair of closely opposed lesions could lead to the formation of double strand breaks that can result in increased lethality and mutagenesis. In vitro processing of closely opposed lesions was studied using double-stranded DNA containing a nick in close proximity opposite to a dihydrouracil. In this study we showed that HU protein, an Escherichia coli DNA-binding protein, has a role in the repair of closely opposed lesions. The repair of dihydrouracil is initiated by E. coli endonuclease III and processed via the base excision repair pathway. HU protein was shown to inhibit the rate of removal of dihydrouracil by endonuclease III only when the DNA substrate contained a nick in close proximity opposite to the dihydrouracil. In contrast, HU protein did not inhibit the subsequent steps of the base excision repair pathway, namely the DNA synthesis and ligation reactions catalyzed by E. coli DNA polymerase and E. coli DNA ligase, respectively. The nick-dependent selective inhibition of endonuclease III activity by HU protein suggests that HU could play a role in reducing the formation of double strand breaks in E. coli.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Type VIII