MilliporeSigma
Rho-associated kinase of chicken gizzard smooth muscle.
The Journal of biological chemistry (1999-01-28)
J Feng, M Ito, Y Kureishi, K Ichikawa, M Amano, N Isaka, K Okawa, A Iwamatsu, K Kaibuchi, D J Hartshorne, T Nakano
ABSTRACT
Rho-associated kinase (Rho-kinase) from chicken gizzard smooth muscle was purified to apparent homogeneity (160 kDa on SDS-polyacrylamide gel electrophoresis) and identified as the ROKalpha isoform. Several substrates were phosphorylated. Rates with myosin phosphatase target subunit 1 (MYPT1), myosin, and the 20-kDa myosin light chain were higher than other substrates. Thiophosphorylation of MYPT1 inhibited myosin phosphatase activity. Phosphorylation of myosin at serine 19 increased actin-activated Mg+-ATPase activity, i.e. similar to myosin light chain kinase. Myosin phosphorylation was increased at higher ionic strengths, possibly by formation of 6 S myosin. Phosphorylation of the isolated light chain and myosin phosphatase was decreased by increasing ionic strength. Rho-kinase was stimulated 1.5-2-fold by guanosine 5'-O-3-(thio)triphosphate.RhoA, whereas limited tryptic hydrolysis caused a 5-6-fold activation, independent of RhoA. Several kinase inhibitors were screened and most effective were Y-27632, staurosporine, and H-89. Several lipids caused slight activation of Rho-kinase, but arachidonic acid (30-50 microM) induced a 5-6-fold activation, independent of RhoA. These results suggest that Rho-kinase of smooth muscle may be involved in the contractile process via phosphorylation of MYPT1 and myosin. Activation by arachidonic acid presents a possible regulatory mechanism for Rho-kinase.
Sigma-Aldrich
ROKα/ROCK-II Protein, active, 10 µg, Active, N-terminal His6-tagged recombinant human ROCK-II residues 11-552, for use in Kinase Assays.
Pricing and availability is not currently available.
Sigma-Aldrich
ROKα/ROCK-II Protein, active, rat, 10 µg, Active, rat, N-terminal, His-tagged, fusion protein corresponding to amino acids 2-543 of rat ROKa/ROCK-II. For use in Kinase Assays.
Pricing and availability is not currently available.