MilliporeSigma
  • SUMOylation of the kainate receptor subunit GluK2 contributes to the activation of the MLK3-JNK3 pathway following kainate stimulation.
SUMOylation of the kainate receptor subunit GluK2 contributes to the activation of the MLK3-JNK3 pathway following kainate stimulation.
FEBS letters (2012-04-10)
Qiu-Ju Zhu, Yan Xu, Cai-Ping Du, Xiao-Yu Hou
ABSTRACT
Protein SUMOylation has been implicated in the pathogenesis of ischemic stroke. However, the underlying mechanisms remain unclear. Here, we found that global brain ischemia evokes a sustained elevation of GluK2 SUMOylation in the rat hippocampal CA1 region. Over-expression of wild-type GluK2, but not SUMOylation-deficient mutant, significantly increased the activity of MLK3 and JNK3 after kainate stimulation. SUMOylation deficiency attenuated the kainate-stimulated interaction between MLK3 and GluK2. In addition, inhibition of kainate-evoked GluK2 endocytosis decreased the activation of MLK3-JNK3 signaling and the binding of MLK3-GluK2 in cultured cortical neurons. These results suggest that the internalization of GluK2 following SUMO modification promotes its binding with MLK3, thereby activating the MLK3-JNK3 pathway, which may be responsible for ischemic neuronal cell death.
Sigma-Aldrich
Anti-GluR6/7 Antibody, clone NL9, rabbit monoclonal, culture supernatant, clone NL904, Upstate®
Pricing and availability is not currently available.