MilliporeSigma
  • Promoter-regulatory region of the major immediate early gene of human cytomegalovirus.
Promoter-regulatory region of the major immediate early gene of human cytomegalovirus.
Proceedings of the National Academy of Sciences of the United States of America (1984-02-01)
D R Thomsen, R M Stenberg, W F Goins, M F Stinski
ABSTRACT
The DNA templates containing immediate early (IE) genes of human cytomegalovirus (CMV) were transcribed in vitro by using a HeLa cell extract. When IE region 1, 2, and 3 were used, transcription was detected qualitatively only from IE region 1. Transcription was detected with DNA representing IE region 2 when the IE region 1 promoter was not present. DNA sequence analysis of the upstream regulatory region of IE region 1 detected two distinct repeats of 19 and 18 nucleotides, both being repeated four times. A putative cruciform structure could form through the surrounding sequences with each 18-nucleotide repeat being located in the unpaired region. The potential secondary structure and the repeat sequences in the regulatory region of IE region 1 are presumably related to the high level of transcription of this IE gene.
Product Description
Millipore
EZview Red Glutathione Affinity Gel
Pricing and availability is not currently available.
Sigma-Aldrich
pBICEP-CMV-3 Expression Vector
Pricing and availability is not currently available.