MilliporeSigma
  • Acetyl Coenzyme A Acetyltransferase of Rhizobium sp. (Cicer) Strain CC 1192.
Acetyl Coenzyme A Acetyltransferase of Rhizobium sp. (Cicer) Strain CC 1192.
Applied and environmental microbiology (2006-03-15)
S A Kim, L Copeland
ABSTRACT
To investigate why Rhizobium sp. (Cicer) strain CC 1192 cells accumulate poly-R-3-hydroxybutyrate in the free-living state but not as bacteroids in nodules on chickpea (Cicer arietinum L.) plants, we have examined the kinetic properties of acetyl coenzyme A (acetyl-CoA) acetyltransferase (also known as acetoacetyl-CoA thiolase and 3-ketothiolase [EC 2.3.1.9]) from both types of cells. The enzyme had a native molecular mass of 180 (plusmn) 4 kDa, and the subunit molecular mass was 44 (plusmn) 1 kDa. The seven amino acids from the N terminus were Lys-Ala-Ser-Ile-Val-Ile-Ala. Thiolysis and condensation activity of the enzyme from free-living CC 1192 cells were optimal at pHs 7.8 and 8.1, respectively. The relationship between substrate concentrations and initial velocity for the thiolysis reaction were hyperbolic and gave K(infm) values for acetoacetyl-CoA and CoA of 42 and 56 (mu)M, respectively. The maximum velocity in the condensation direction was approximately 10% of that of the thiolysis reaction. With highly purified preparations of the enzyme, a value of approximately 1 mM was determined for the apparent K(infm) for acetyl-CoA. However, with partially purified enzyme preparations or when N-ethylmaleimide was included in reaction mixtures the apparent K(infm) for acetyl-CoA was close to 0.3 mM. In the condensation direction, CoA was a potent linear competitive inhibitor with an inhibition constant of 11 (mu)M. The much higher affinity of the enzyme for the product CoA than the substrate acetyl-CoA could have significance in view of metabolic differences between bacteroid and free-living cells of CC 1192. We propose that in free-living CC 1192 cells, the acetyl-CoA/CoA ratio reaches a value that allows condensation activity of acetyl-CoA acetyltransferase, but that in CC 1192 bacteroids, the ratio is poised so that the formation of acetoacetyl-CoA is not favored.