MilliporeSigma
  • NMR studies of a ferredoxin from Haloferax mediterranei and its physiological role in nitrate assimilatory pathway.
NMR studies of a ferredoxin from Haloferax mediterranei and its physiological role in nitrate assimilatory pathway.
Biochimica et biophysica acta (2003-09-06)
Rosa M Martínez-Espinosa, Frutos C Marhuenda-Egea, Antonio Donaire, María José Bonete
ABSTRACT
Haloferax mediterranei is a halophilic archaeon that can grow in aerobic conditions with nitrate as sole nitrogen source. The electron donor in the aerobic nitrate reduction to ammonium was a ferredoxin. This ferredoxin has been purified and characterised. Air-oxidized H. mediterranei ferredoxin has a UV-visible absorption spectra typical of 2Fe-type ferredoxins with an A420/A280 of 0.21. The nuclear magnetic resonance (NMR) spectra of the ferredoxin showed similarity to those of ferredoxins from plant and bacteria, containing a [2Fe-2S] cluster. The physiological function of ferredoxin might be to serve as an electron donor for nitrate reduction to ammonium by assimilatory nitrate (EC 1.6.6.2) and nitrite reductases (EC 1.7.7.1). The apparent molecular weight (Mr) of the ferredoxin was estimated to be 21 kDa on SDS-polyacrylamide gel electrophoresis (SDS-PAGE).
Sigma-Aldrich
Ferredoxin from Spinacia oleracea (spinach), lyophilized powder, Ferredoxin ≥15 wt. %
Pricing and availability is not currently available.
Sigma-Aldrich
Ferredoxin from Spinacia oleracea (spinach), buffered aqueous solution
Pricing and availability is not currently available.