MilliporeSigma
  • A Catalytic Cross-Olefination of Diazo Compounds with Sulfoxonium Ylides.
A Catalytic Cross-Olefination of Diazo Compounds with Sulfoxonium Ylides.
Angewandte Chemie (International ed. in English) (2018-09-29)
James D Neuhaus, Adriano Bauer, Alexandre Pinto, Nuno Maulide
ABSTRACT
A ruthenium-catalysed cross-olefination of diazo compounds and sulfoxonium ylides is presented. Our reaction design exploits the intrinsic difference in reactivity of diazo compounds and sulfoxonium ylides as both carbene precursors and nucleophiles, which results in a highly selective reaction.