MilliporeSigma
  • Identification of two genes required for heptadecane production in a N2-fixing cyanobacterium Anabaena sp. strain PCC 7120.
Identification of two genes required for heptadecane production in a N2-fixing cyanobacterium Anabaena sp. strain PCC 7120.
AMB Express (2018-10-15)
Jaimie Gibbons, Liping Gu, Huilan Zhu, William Gibbons, Ruanbao Zhou
ABSTRACT
Cyanobacteria photosynthetically produce long-chain hydrocarbons, which are considered as infrastructure-compatible biofuels. However, native cyanobacteria do not produce these hydrocarbons at sufficient rates or yields to warrant commercial deployment. This research sought to identify specific genes required for photosynthetic production of alkanes to enable future metabolic engineering for commercially viable production of alkanes. The two putative genes (alr5283 and alr5284) required for long-chain hydrocarbon production in Anabaena sp. PCC 7120 were knocked out through a double crossover approach. The knockout mutant abolished the production of heptadecane (C17H36). The mutant is able to be complemented by a plasmid bearing the two genes along with their native promoters only. The complemented mutant restored photosynthetic production of heptadecane. This combined genetic and metabolite (alkanes) profiling approach may be broadly applicable to characterization of knockout mutants, using N2-fixing cyanobacteria as a cellular factory driven by solar energy to produce a wide range of commodity chemicals and drop-in-fuels from atmospheric gases (CO2 and N2 gas) and mineralized water.
Sigma-Aldrich
1-Pentadecene, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Heptadecene, 98%
Pricing and availability is not currently available.