MilliporeSigma
GPR17 mediates ischemia-like neuronal injury via microglial activation.
International journal of molecular medicine (2018-09-19)
Bing Zhao, Hao Wang, Cai-Xia Li, Sheng-Wen Song, San-Hua Fang, Er-Qing Wei, Qiao-Juan Shi
ABSTRACT
GPR17 is a G (i)-coupled dual receptor, linked to P2Y and CysLT receptors stimulated by uracil nucleotides and cysteinyl leukotrienes, respectively. Recent evidence has demonstrated that GPR17 inhibition ameliorates the progression of cerebral ischemic injury by regulating neuronal death and microglial activation. The present study aimed to assess the detailed regulatory roles of this receptor in oxygen‑glucose deprivation/recovery (OGD/R)‑induced ischemia‑like injury in vitro and explore the underlying mechanism. The results demonstrated that OGD/R induced ischemic neuronal injury and microglial activation, including enhanced phagocytosis and increased inflammatory cytokine release in neuron‑glial mixed cultures of cortical cells. GPR17 upregulation during OGD/R was spatially and temporally correlated with neuronal injury and microglial activation. In addition, GPR17 knockdown inhibited OGD/R‑induced responses in neuron‑glial mixed cultures. GPR17 knockdown also attenuated cell injury induced by the agonist leukotriene D4 (LTD4) or uridine 5'‑diphosphate (UDP) in neuron‑glial mixed cultures. However, GPR17 knockdown did not affect OGD/R‑induced ischemic neuronal injury in primary cultures of neurons. In primary astrocyte cultures, neither GPR17 nor OGD/R induced injury. By contrast, GPR17 knockdown ameliorated OGD/R‑induced microglial activation, boosting phagocytosis and inflammatory cytokine release in primary microglia cultures. Finally, the results demonstrated that the conditioned medium of microglia pretreated with OGD/R induced neuronal death, and the neuronal injury was significantly inhibited by GPR17 knockdown. These findings suggested that GPR17 may mediate ischemia‑like neuronal injury and microglial activation in vitro; however, the protective effects on ischemic neuronal injury might depend upon microglial activation. Whether GPR17 regulates neuronal injury mediated by oligodendrocyte linkage remains to be investigated.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human GPR17
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed, 1.5 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) FITC Conjugated, 2 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Leukotriene D4, ~50 μg/mL (in methanol/ammonium acetate buffer, 70:30, pH 5.6), ≥97%