MilliporeSigma
Reduction of amine N-oxides by diboron reagents.
The Journal of organic chemistry (2011-08-05)
Hari Prasad Kokatla, Paul F Thomson, Suyeal Bae, Venkata Ramana Doddi, Mahesh K Lakshman
ABSTRACT
Facile reduction of alkylamino-, anilino-, and pyridyl-N-oxides can be achieved via the use of diboron reagents, predominantly bis(pinacolato)- and in some cases bis(catecholato)diboron [(pinB)(2) and (catB)(2), respectively]. Reductions occur upon simply mixing the amine N-oxide and the diboron reagent in a suitable solvent, at a suitable temperature. Extremely fast reductions of alkylamino- and anilino-N-oxides occur, whereas pyridyl-N-oxides undergo slower reduction. The reaction is tolerant of a variety of functionalities such as hydroxyl, thiol, and cyano groups, as well as halogens. Notably, a sensitive nucleoside N-oxide has also been reduced efficiently. The different rates with which alkylamino- and pyridyl-N-oxides are reduced has been used to perform stepwise reduction of the N,N'-dioxide of (S)-(-)-nicotine. Because it was observed that (pinB)(2) was unaffected by the water of hydration in amine oxides, the feasibility of using water as solvent was evaluated. These reactions also proceeded exceptionally well, giving high product yields. In constrast to the reactions with (pinB)(2), triethylborane reduced alkylamino-N-oxides, but pyridine N-oxide did not undergo efficient reduction even at elevated temperature. Finally, the mechanism of the reductive process by (pinB)(2) has been probed by (1)H and (11)B NMR.