MilliporeSigma
  • Rho A Regulates Epidermal Growth Factor-Induced Human Osteosarcoma MG63 Cell Migration.
Rho A Regulates Epidermal Growth Factor-Induced Human Osteosarcoma MG63 Cell Migration.
International journal of molecular sciences (2018-05-13)
Jinyang Wang, Lei Zhang, Rongmei Qu, Lin Zhang, Wenhua Huang
ABSTRACT
Osteosarcoma, the most common primary bone tumor, occurs most frequently in children and adolescents and has a 5-year survival rate, which is unsatisfactory. As epidermal growth factor receptor (EGFR) positively correlates with TNM (tumor-node-metastasis) stage in osteosarcoma, EGFR may play an important role in its progression. The purpose of this study was to explore potential mechanisms underlying this correlation. We found that EGF promotes MG63 cell migration and invasion as well as stress fiber formation via Rho A activation and that these effects can be reversed by inhibiting Rho A expression. In addition, molecules downstream of Rho A, including ROCK1, LIMK2, and Cofilin, are activated by EGF in MG63 cells, leading to actin stress fiber formation and cell migration. Moreover, inhibition of ROCK1, LIMK2, or Cofilin in MG63 cells using known inhibitors or short hairpin RNA (shRNA) prevents actin stress fiber formation and cell migration. Thus, we conclude that Rho A/ROCK1/LIMK2/Cofilin signaling mediates actin microfilament formation in MG63 cells upon EGFR activation. This novel pathway provides a promising target for preventing osteosarcoma progression and for treating this cancer.
Sigma-Aldrich
Anti-phospho-LIMK2 (pThr505) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.