MilliporeSigma
  • Genome-wide regulon and crystal structure of BlaI (Rv1846c) from Mycobacterium tuberculosis.
Genome-wide regulon and crystal structure of BlaI (Rv1846c) from Mycobacterium tuberculosis.
Molecular microbiology (2009-01-22)
Claudia Sala, Ahmed Haouz, Frederick A Saul, Isabelle Miras, Ida Rosenkrands, Pedro M Alzari, Stewart T Cole
ABSTRACT
Comparative genomics with Staphylococcus aureus suggested the existence of a regulatory system governing beta-lactamase (BlaC) production in Mycobacterium tuberculosis. The crystal structure of Rv1846c, a winged helix regulator of previously unknown function, was solved thus revealing strong similarity to the BlaI and MecI repressors of S. aureus, which both respond to beta-lactam treatment. Using chromatin immunoprecipitation and hybridization to microarrays (ChIP-on-chip), the Rv1846c regulon was shown to comprise five separate genomic loci. Two of these mediate responses and resistance to beta-lactam antibiotics (rv1845c, rv1846c-rv1847; blaC-sigC); two encode membrane proteins of unknown function (rv1456c, rv3921c) while the last codes for ATP synthase (rv1303-atpBEFHAGDC-rv1312). The ChIP-on-chip findings were confirmed independently using electrophoretic mobility shift assays, DNAse footprinting and transcript analysis leading to Rv1846c being renamed BlaI. When cells were treated with beta-lactams, BlaI was released from its operator sites causing derepression of the regulon and upregulation of ATP synthase transcription. The existence of a potential regulatory loop between cell wall integrity and ATP production was previously unknown.
Sigma-Aldrich
Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.