MilliporeSigma
  • Development of a novel immunoproteasome digestion assay for synthetic long peptide vaccine design.
Development of a novel immunoproteasome digestion assay for synthetic long peptide vaccine design.
PloS one (2018-07-04)
Hiroshi Wada, Atsushi Shimizu, Toshihiro Osada, Yuki Tanaka, Satoshi Fukaya, Eiji Sasaki
ABSTRACT
Recently, many autologous tumor antigens have been examined for their potential use in cancer immunotherapy. However, the success of cancer vaccines in clinical trials has been limited, partly because of the limitations of using single, short peptides in most attempts. With this in mind, we aimed to develop multivalent synthetic long peptide (SLP) vaccines containing multiple cytotoxic T-lymphocyte (CTL) epitopes. However, to confirm whether a multivalent vaccine can induce an individual epitope-specific CTL, the only viable screening strategies currently available are interferon-gamma (IFN-μ enzyme-linked immunospot (ELISPOT) assays using human peripheral blood mononuclear cells, or expensive human leukocyte antigen (HLA)-expressing mice. In this report, we evaluated the use of our developed murine-20S immunoproteasome (i20S) digestion assay, and found that it could predict the results of IFN-μ ELISPOT assays. Importantly, the murine-i20S digestion assay not only predicted CTL induction, but also antitumor activity in an HLA-expressing mouse model. We conclude that the murine-i20S digestion assay is an extremely useful tool for the development of "all functional" multivalent SLP vaccines.
Sigma-Aldrich
HBTU, ≥98.0% (T)