MilliporeSigma
  • APOA-1Milano muteins, orally delivered via genetically modified rice, show anti-atherogenic and anti-inflammatory properties in vitro and in Apoe-/- atherosclerotic mice.
APOA-1Milano muteins, orally delivered via genetically modified rice, show anti-atherogenic and anti-inflammatory properties in vitro and in Apoe-/- atherosclerotic mice.
International journal of cardiology (2018-06-17)
Gabriele Romano, Serena Reggi, Barbara Kutryb-Zajac, Amanda Facoetti, Elisa Chisci, Mariateresa Pettinato, Maria Rita Giuffrè, Federica Vecchio, Silvia Leoni, Marco De Giorgi, Federica Avezza, Massimiliano Cadamuro, Luca Crippa, Biagio Eugenio Leone, Marialuisa Lavitrano, Ilaria Rivolta, Donatella Barisani, Ryszard Tomasz Smolenski, Roberto Giovannoni
ABSTRACT
Atherosclerosis is a slowly progressing, chronic multifactorial disease characterized by the accumulation of lipids, inflammatory cells, and fibrous tissue that drives to the formation of asymmetric focal thickenings in the tunica intima of large and mid-sized arteries. Despite the high therapeutic potential of ApoA-1 proteins, the purification and delivery into the disordered organisms of these drugs is still limited by low efficiency in these processes. We report here a novel production and delivery system of anti-atherogenic APOA-1Milano muteins (APOA-1M) by means of genetically modified rice plants. APOA-1M, delivered as protein extracts from transgenic rice seeds, significantly reduced macrophage activation and foam cell formation in vitro in oxLDL-loaded THP-1 model. The APOA-1M delivery method and therapeutic efficacy was tested in healthy mice and in Apoe-/- mice fed with high cholesterol diet (Western Diet, WD). APOA-1M rice milk significantly reduced atherosclerotic plaque size and lipids composition in aortic sinus and aortic arch of WD-fed Apoe-/- mice as compared to wild type rice milk-treated, WD-fed Apoe-/- mice. APOA-1M rice milk also significantly reduced macrophage number in liver of WD-fed Apoe-/- mice as compared to WT rice milk treated mice. The delivery of therapeutic APOA-1M full length proteins via oral administration of rice seeds protein extracts (the 'rice milk') to the disordered organism, without any need of purification, might overcome the main APOA1-based therapies' limitations and improve the use of this molecules as therapeutic agents for cardiovascular patients.
Sigma-Aldrich
Apolipoprotein A-I from human plasma, ≥85% (SDS-PAGE), buffered aqueous solution