MilliporeSigma
  • Microwave-assisted synthesis of thymine-functionalized graphitic carbon nitride quantum dots as a fluorescent nanoprobe for mercury(II).
Microwave-assisted synthesis of thymine-functionalized graphitic carbon nitride quantum dots as a fluorescent nanoprobe for mercury(II).
Mikrochimica acta (2018-09-17)
Ojodomo J Achadu, Neerish Revaprasadu
ABSTRACT
A microwave-assisted hydrothermal method was employed to prepare thymine-modified graphitic carbon nitride quantum dots (T-gCNQDs) which are shown to be a novel fluorescent nanoprobe for Hg(II). They exhibit excellent optical properties (blue emission with a fluorescence quantum yield of 46%) and water solubility. The incorporation of thymine into the gCNQDs results in an enhancement in photoluminescence properties. It is found that fluorescence, best measured at excitation/emission wavelengths of 350/445 nm, is much more strongly quenched by Hg(II) compared to the thymine-free nanoprobe. The quenching is highly selective even in the presence other metal ions. This is ascribed to the formation of T-Hg(II)-T base complexes. Fluorescence drops linearly in the 1.0 to 500 nM Hg(II) concentration range, and the limit of detection is 0.15 nM. The method was applied to the determination of Hg(II) in spiked samples of tap and pond water. Recoveries were found to be >95%, thus demonstrating the practical applicability of the assay. Graphical abstract A microwave-assisted hydrothermal route was employed to prepare thymine-functionalized graphitic carbon nitride QDs (T-gCNQDs). A selective fluorescence quenching mechanism occurred between T-gCNQDs and Hg(II) due to thymine functionalization. T-gCNQDs was utilized to detect Hg(II) in real samples.
Sigma-Aldrich
Diaminomaleonitrile, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Thymine-1-acetic acid, 98%
Pricing and availability is not currently available.