MilliporeSigma
  • DNA-Based Sensor for the Detection of an Organophosphorus Pesticide: Profenofos.
DNA-Based Sensor for the Detection of an Organophosphorus Pesticide: Profenofos.
Sensors (Basel, Switzerland) (2018-06-27)
Giulia Selvolini, Ioana Băjan, Oana Hosu, Cecilia Cristea, Robert Săndulescu, Giovanna Marrazza
ABSTRACT
In this work, we propose an electrochemical DNA aptasensor for the detection of profenofos, an organophosphorus pesticide, based on a competitive format and disposable graphite screen-printed electrodes (GSPEs). A thiol-tethered DNA capture probe, which results to be complementary to the chosen aptamer sequence, was immobilised on gold nanoparticles/polyaniline composite film-modified electrodes (AuNPs/PANI/GSPE). Different profenofos solutions containing a fixed amount of the biotinylated DNA aptamer were dropped onto the realized aptasensors. The hybridisation reaction was measured using a streptavidin-alkaline phosphatase enzyme conjugate, which catalyses the hydrolysis of 1-naphthyl -phosphate. The 1-naphtol enzymatic product was detected by means of differential pulse voltammetry (DPV). The aptasensor showed itself to work as a signal off sensor, according to the competitive format used. A dose response curve was obtained between 0.10 μM and 10 μM with a detection limit of 0.27 μM.
Sigma-Aldrich
1-Naphthyl phosphate, 99%