MilliporeSigma
  • Engineered bidirectional promoters enable rapid multi-gene co-expression optimization.
Engineered bidirectional promoters enable rapid multi-gene co-expression optimization.
Nature communications (2018-09-06)
Thomas Vogl, Thomas Kickenweiz, Julia Pitzer, Lukas Sturmberger, Astrid Weninger, Bradley W Biggs, Eva-Maria Köhler, Armin Baumschlager, Jasmin Elgin Fischer, Patrick Hyden, Marlies Wagner, Martina Baumann, Nicole Borth, Martina Geier, Parayil Kumaran Ajikumar, Anton Glieder
ABSTRACT
Numerous synthetic biology endeavors require well-tuned co-expression of functional components for success. Classically, monodirectional promoters (MDPs) have been used for such applications, but MDPs are limited in terms of multi-gene co-expression capabilities. Consequently, there is a pressing need for new tools with improved flexibility in terms of genetic circuit design, metabolic pathway assembly, and optimization. Here, motivated by nature's use of bidirectional promoters (BDPs) as a solution for efficient gene co-expression, we generate a library of 168 synthetic BDPs in the yeast Komagataella phaffii (syn. Pichia pastoris), leveraging naturally occurring BDPs as a parts repository. This library of synthetic BDPs allows for rapid screening of diverse expression profiles and ratios to optimize gene co-expression, including for metabolic pathways (taxadiene, β-carotene). The modular design strategies applied for creating the BDP library could be relevant in other eukaryotic hosts, enabling a myriad of metabolic engineering and synthetic biology applications.
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)