MilliporeSigma
  • Pd-Catalyzed Decarbonylative C-H Coupling of Azoles and Aromatic Esters.
Pd-Catalyzed Decarbonylative C-H Coupling of Azoles and Aromatic Esters.
Chemistry, an Asian journal (2018-05-03)
Kaoru Matsushita, Ryosuke Takise, Tomoya Hisada, Shin Suzuki, Ryota Isshiki, Kenichiro Itami, Kei Muto, Junichiro Yamaguchi
ABSTRACT
A decarbonylative C-H coupling of azoles and aromatic esters by palladium catalysis is described. Our previously reported Ni-catalyzed C-H coupling of azoles and aromatic esters has a significant drawback regarding the substrate scope. Herein, we employ palladium catalysis instead of nickel, resulting in a broader substrate scope in terms of azoles and aromatic esters.
Sigma-Aldrich
Phenyl benzoate, 99%
Pricing and availability is not currently available.