MilliporeSigma
2-Naphthol Levels and Allergic Disorders in Children.
International journal of environmental research and public health (2018-07-11)
Tien-Jen Lin, Yueliang Leon Guo, Jiin-Chyr Hsu, I-Jen Wang
ABSTRACT
The measurement of polycyclic aromatic hydrocarbons (PAH) in ambient air is quite difficult to perform. Using urine biomarkers of PAH such as 2-naphthol is one approach to this problem. This study explored the association between urine 2-naphthol levels and allergic diseases. The associations between 2-naphthol levels and oxidative stress biomarkers for the possible disease pathogenesis were also investigated. A total of 453 kindergarten children from the (Childhood Environment and Allergic Diseases Study) CEAS cohort with urine samples were recruited. Urine 2-naphthol levels were measured by high-performance liquid chromatography mass spectrometry (HPLC-MS/MS) and markers of oxidative stress (8OHdG) were measured by enzyme-linked immunosorbent assays (ELISA). Information on environmental risk factors and allergic diseases were also collected. The association between 2-naphthol levels, 8OHdG levels, IgE, and allergic diseases were evaluated by multivariate linear regression and logistic regression. Levels of 2-naphthol were positively correlated with 8OHdG levels. A one ln-unit increase in the 2-naphthol level was positively associated to 8OHdG levels (per ln-unit: β = 100.61, p < 0.001). When dividing 2-naphthol levels into quartiles, asthma was significantly associated with 2-naphthol levels at a concentration of >1.60 ng/mL (adjusted OR: 3.14, 95% CI 1.34⁻7.35). Urine 2-naphthol levels are associated with markers of oxidative stress and the risk of allergic diseases in young children.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL