MilliporeSigma
  • Purification of chondroitinase B and chondroitinase C using glycosaminoglycan-bound AH-Sepharose 4B.
Purification of chondroitinase B and chondroitinase C using glycosaminoglycan-bound AH-Sepharose 4B.
Carbohydrate research (1979-05-01)
N Ototani, Z Yosizawa
PMID427837
ABSTRACT
Chondroitinase B and chondroitinase C were separated from an extract of Flavobacterium heparinum induced with chondroitin 6-sulfate by using column chromatography on hydroxylapatite. Chondroitinase C was eluted together with the activities of hyaluronidase, delta4,5glycosiduronase, and sulfatase. The latter two activities were eliminated exclusively by passing the crude chondroitinase C fraction through a phosphono-cellulose column pre-equilibrated with 0.07M sodium phosphate buffer (pH 6.8). Chondroitinase C was then purified by affinity chromatography using dermatan sulfate-bound AH-Sepharose 4B coated with the same glycosaminoglycan. Purification of the enzyme was achieved 18-fold and in 73% yield. On the other hand, the activities of delta4,5glycosiduronase and sulfatase were decreased to 50 and 60%, respectively, as compared with those in the crude chondroitinase B fraction, after passing the fraction through a column of phosphono-cellulose pre-equilibrated with 0.1M sodium phosphate buffer (pH 6.8). The remaining activities of these two enzymes were then eliminated from chondroitinase B by affinity chromatography with heparin-bound AH-Sepharose 4B coated with dermatan sulfate. In the affinity chromatography used in the present study, non-covalent coating of the glycosaminoglycan-bound (covalently) AH-Sepharose 4B with the same or another glycosaminoglycan was found to be important.
Sigma-Aldrich
Chondroitinase C from Flavobacterium heparinum, lyophilized powder, ≥200 units/mg solid
Pricing and availability is not currently available.