MilliporeSigma
  • Identification of 4-hydroxy-N-desmethyltamoxifen as a metabolite of tamoxifen in human bile.
Identification of 4-hydroxy-N-desmethyltamoxifen as a metabolite of tamoxifen in human bile.
Cancer research (1988-04-15)
E A Lien, E Solheim, S Kvinnsland, P M Ueland
ABSTRACT
The occurrence of tamoxifen metabolites in bile was investigated in a 57-year-old female patient receiving chronic treatment with tamoxifen. In bile treated with beta-glucuronidase, two major peaks were detected using a chromatographic system developed for the quantitation of tamoxifen metabolites in human serum. One sharp peak coeluted with 4-hydroxy-tamoxifen whereas a second broad peak eluted slightly ahead of tamoxifen and was separated from all major serum metabolites. This latter peak was identified as the cis (about 30%) and trans (about 70%) isomers of 4-hydroxy-N-desmethyltamoxifen. The identification was based on (a) coelution with authentic standard on reversed-phase chromatography and formation of fluorescent material after photoactivation, (b) a molecular ion (M + 1)+ of 374 m/z as determined with liquid chromatography-mass spectrometry, and (c) a fragmentogram identical to that of the authentic standard, as obtained by gas chromatography-mass spectrometry.
Sigma-Aldrich
(Z)-Endoxifen, ≥98% (HPLC)
Pricing and availability is not currently available.