MilliporeSigma
Targeting Plasmodium PI(4)K to eliminate malaria.
Nature (2013-11-29)
Case W McNamara, Marcus Cs Lee, Chek Shik Lim, Siau Hoi Lim, Jason Roland, Oliver Simon, Bryan Ks Yeung, Arnab K Chatterjee, Susan L McCormack, Micah J Manary, Anne-Marie Zeeman, Koen J Dechering, Tr Santha Kumar, Philipp P Henrich, Kerstin Gagaring, Maureen Ibanez, Nobutaka Kato, Kelli L Kuhen, Christoph Fischli, Advait Nagle, Matthias Rottmann, David M Plouffe, Badry Bursulaya, Stephan Meister, Lucia Rameh, Joerg Trappe, Dorothea Haasen, Martijn Timmerman, Robert W Sauerwein, Rossarin Suwanarusk, Bruce Russell, Laurent Renia, Francois Nosten, David C Tully, Clemens Hm Kocken, Richard J Glynne, Christophe Bodenreider, David A Fidock, Thierry T Diagana, Elizabeth A Winzeler
ABSTRACT
Achieving the goal of malaria elimination will depend on targeting Plasmodium pathways essential across all life stages. Here we identify a lipid kinase, phosphatidylinositol-4-OH kinase (PI(4)K), as the target of imidazopyrazines, a new antimalarial compound class that inhibits the intracellular development of multiple Plasmodium species at each stage of infection in the vertebrate host. Imidazopyrazines demonstrate potent preventive, therapeutic, and transmission-blocking activity in rodent malaria models, are active against blood-stage field isolates of the major human pathogens P. falciparum and P. vivax, and inhibit liver-stage hypnozoites in the simian parasite P. cynomolgi. We show that imidazopyrazines exert their effect through inhibitory interaction with the ATP-binding pocket of PI(4)K, altering the intracellular distribution of phosphatidylinositol-4-phosphate. Collectively, our data define PI(4)K as a key Plasmodium vulnerability, opening up new avenues of target-based discovery to identify drugs with an ideal activity profile for the prevention, treatment and elimination of malaria.
Sigma-Aldrich
KDU691, ≥98% (HPLC)
Pricing and availability is not currently available.