MilliporeSigma
  • RSK-B, a novel ribosomal S6 kinase family member, is a CREB kinase under dominant control of p38alpha mitogen-activated protein kinase (p38alphaMAPK).
RSK-B, a novel ribosomal S6 kinase family member, is a CREB kinase under dominant control of p38alpha mitogen-activated protein kinase (p38alphaMAPK).
The Journal of biological chemistry (1998-10-29)
B Pierrat, J S Correia, J L Mary, M Tomás-Zuber, W Lesslauer
ABSTRACT
A novel ribosomal S6 kinase (RSK) family member, RSK-B, was identified in a p38alphaMAPK-baited intracellular interaction screen. RSK-B presents two catalytic domains typical for the RSK family. The protein kinase C-like N-terminal and the calcium/calmodulin kinase-like C-terminal domains both contain conserved ATP-binding and activation consensus sequences. RSK-B is a p38alphaMAPK substrate, and activated by p38alphaMAPK and, more weakly, by ERK1. RSK-B phosphorylates the cAMP response element-binding protein (CREB) and c-Fos peptides. In intracellular assays, RSK-B drives cAMP response element- and AP1-dependent reporter expression. RSK-B locates to the cell nucleus and co-translocates p38alphaMAPK. In conclusion, RSK-B is a novel CREB kinase under dominant p38alphaMAPK control, also phosphorylating additional substrates.
Sigma-Aldrich
MSK2, active, GST tagged from mouse, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.