MilliporeSigma
  • Quantification of microplastic mass and removal rates at wastewater treatment plants applying Focal Plane Array (FPA)-based Fourier Transform Infrared (FT-IR) imaging.
Quantification of microplastic mass and removal rates at wastewater treatment plants applying Focal Plane Array (FPA)-based Fourier Transform Infrared (FT-IR) imaging.
Water research (2018-05-29)
Márta Simon, Nikki van Alst, Jes Vollertsen
ABSTRACT
This paper presents a method for microplastic (MP) mass quantification using a Focal Plane Array-based Fourier Transform Infrared imaging technique. It discusses the issue that particle number is not a conserved base quantity and hence less suited than mass to compare independent studies on MP in the environment. It concludes that MP mass should be included when quantifying MP pollution in the environment, supplementing the conventional approach of reporting particle numbers. Applying mass as the unit of MP measurement, the paper presents data showing that Danish wastewater treatment plants discharge around 3 t/year of MP in the size range 10-500 μm. This value corresponds to an annual per capita emission from these plants of 0.56 g MP/(capita year). The distribution of polymer types by mass and particle number differed because the size of MP particles of the different material types varied.
Sigma-Aldrich
Micro particles based on polystyrene, dark red, size: 100 μm