MilliporeSigma
  • In situ ionic liquid dispersive liquid-liquid microextraction coupled to gas chromatography-mass spectrometry for the determination of organophosphorus pesticides.
In situ ionic liquid dispersive liquid-liquid microextraction coupled to gas chromatography-mass spectrometry for the determination of organophosphorus pesticides.
Journal of chromatography. A (2018-01-02)
J I Cacho, N Campillo, P Viñas, M Hernández-Córdoba
ABSTRACT
Nine organophosphorus pesticides (OPPs) were determined in environmental waters from different origins using in situ ionic liquid dispersive liquid microextraction (IL-DLLME). This preconcentration technique was coupled to gas chromatography-mass spectrometry (GC-MS) using microvial insert thermal desorption, an approach that uses a thermal desorption injector as sample introduction system. The parameters affecting both the microextraction and sample injection steps were optimized. The proposed method showed good precision, with RSD values ranging from 4.1 to 9.7%, accuracy with recoveries in the 85-118% range, and sensitivity with DLs ranging from 5 to 16 ng L-1.
Product Description
Supelco
Phorate, PESTANAL®, analytical standard
Supelco
Sulfotep, PESTANAL®, analytical standard