MilliporeSigma
  • Novel 3D Hybrid Nanofiber Aerogels Coupled with BMP-2 Peptides for Cranial Bone Regeneration.
Novel 3D Hybrid Nanofiber Aerogels Coupled with BMP-2 Peptides for Cranial Bone Regeneration.
Advanced healthcare materials (2018-03-03)
Lin Weng, Sunil Kumar Boda, Hongjun Wang, Matthew J Teusink, Franklin D Shuler, Jingwei Xie
ABSTRACT
An ideal synthetic bone graft is a combination of the porous and nanofibrous structure presented by natural bone tissue as well as osteoinductive biochemical factors such as bone morphogenetic protein 2 (BMP-2). In this work, ultralight 3D hybrid nanofiber aerogels composed of electrospun PLGA-collagen-gelatin and Sr-Cu codoped bioactive glass fibers with incorporation of heptaglutamate E7 domain specific BMP-2 peptides have been developed and evaluated for their potential in cranial bone defect healing. The nanofiber aerogels are surgically implanted into 8 mm × 1 mm (diameter × thickness) critical-sized defects created in rat calvariae. A sustained release of E7-BMP-2 peptide from the degradable hybrid aerogels significantly enhances bone healing and defect closure over 8 weeks in comparison to unfilled defects. Histomorphometry and X-ray microcomputed tomography (µ-CT) analysis reveal greater bone volume and bone formation area in case of the E7-BMP-2 peptide loaded hybrid nanofiber aerogels. Further, histopathology data divulged a near complete nanofiber aerogel degradation along with enhanced vascularization of the regenerated tissue. Together, this study for the first time demonstrates the fabrication of 3D hybrid nanofiber aerogels from 2D electrospun fibers and their loading with therapeutic osteoinductive BMP-2 mimicking peptide for cranial bone tissue regeneration.
Sigma-Aldrich
ε-Caprolactone, 97%
Pricing and availability is not currently available.