MilliporeSigma
  • Centrifugation-free extraction of circulating nucleic acids using immiscible liquid under vacuum pressure.
Centrifugation-free extraction of circulating nucleic acids using immiscible liquid under vacuum pressure.
Scientific reports (2018-04-05)
Hoyoon Lee, Wonhwi Na, Chanhee Park, Kyong Hwa Park, Sehyun Shin
ABSTRACT
Extraction of cell-free DNA (cfDNA), which exists at an extremely low concentration in plasma, is a critical process for either targeted-sensing or massive sequencing of DNAs. However, such small amount of DNA cannot be fully obtained without high-speed centrifugation (<20,000 g). Here, we developed a centrifugation-free cfDNA extraction method and system that utilizes an immiscible solvent under single low vacuum pressure throughout the entire process. It has been named Pressure and Immiscibility-Based EXtraction (PIBEX). The amounts of extracted cfDNA by PIBEX were compared with those extracted by the conventional gold standards such as QIAGEN using quantitative PCR (qPCR). The PIBEX system showed equal performance regarding extraction amount and efficiency compared to the existing method. Because the PIBEX eliminates the troublous and repetitive centrifugation processes in DNA extraction, it can be further utilized in microfluidic-sample preparation systems for circulating nucleic acids, which would lead to an integrated sample-to-answer system in liquid biopsies.
Sigma-Aldrich
Mineral oil, heavy
Pricing and availability is not currently available.