MilliporeSigma
  • The hip fracture surgery in elderly patients (HIPELD) study to evaluate xenon anaesthesia for the prevention of postoperative delirium: a multicentre, randomized clinical trial.
The hip fracture surgery in elderly patients (HIPELD) study to evaluate xenon anaesthesia for the prevention of postoperative delirium: a multicentre, randomized clinical trial.
British journal of anaesthesia (2018-02-06)
M Coburn, R D Sanders, M Maze, M-L Nguyên-Pascal, S Rex, B Garrigues, J A Carbonell, M L Garcia-Perez, A Stevanovic, P Kienbaum, M Neukirchen, M S Schaefer, B Borghi, H van Oven, A Tognù, L Al Tmimi, L Eyrolle, O Langeron, X Capdevila, G M Arnold, M Schaller, R Rossaint
ABSTRACT
Postoperative delirium occurs frequently in elderly hip fracture surgery patients and is associated with poorer overall outcomes. Because xenon anaesthesia has neuroprotective properties, we evaluated its effect on the incidence of delirium and other outcomes after hip fracture surgery. This was a phase II, multicentre, randomized, double-blind, parallel-group, controlled clinical trial conducted in hospitals in six European countries (September 2010 to October 2014). Elderly (≥75yr-old) and mentally functional hip fracture patients were randomly assigned 1:1 to receive either xenon- or sevoflurane-based general anaesthesia during surgery. The primary outcome was postoperative delirium diagnosed through postoperative day 4. Secondary outcomes were delirium diagnosed anytime after surgery, postoperative sequential organ failure assessment (SOFA) scores, and adverse events (AEs). Of 256 enrolled patients, 124 were treated with xenon and 132 with sevoflurane. The incidence of delirium with xenon (9.7% [95% CI: 4.5 -14.9]) or with sevoflurane (13.6% [95% CI: 7.8 -19.5]) were not significantly different (P=0.33). Overall SOFA scores were significantly lower with xenon (least-squares mean difference: -0.33 [95% CI: -0.60 to -0.06]; P=0.017). For xenon and sevoflurane, the incidence of serious AEs and fatal AEs was 8.0% vs 15.9% (P=0.05) and 0% vs 3.8% (P=0.06), respectively. Xenon anaesthesia did not significantly reduce the incidence of postoperative delirium after hip fracture surgery. Nevertheless, exploratory observations concerning postoperative SOFA-scores, serious AEs, and deaths warrant further study of the potential benefits of xenon anaesthesia in elderly hip fracture surgery patients. EudraCT 2009-017153-35; ClinicalTrials.gov NCT01199276.