MilliporeSigma
  • MiR-384 represses tumorigenesis by regulating CDK6 and predicts prognosis of clear cell renal cell carcinoma.
MiR-384 represses tumorigenesis by regulating CDK6 and predicts prognosis of clear cell renal cell carcinoma.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology (2018-07-14)
Lihua Guo, Dayu Wang, Zhiping Zhang
ABSTRACT
Clear cell renal cell carcinoma (ccRCC) accounts for 70-80% of renal cell carcinomas. Various microRNAs (miRs) have been reported to affect the tumorigenesis of ccRCC. However, the role of miR-384 in ccRCC is still unknown. Thus, the purpose of this study was to investigate the function of miR-384 in ccRCC. qRT-PCR or Western blot were employed to examine the expressions of miR-384 and CDK6 in ccRCC tissues or cell lines. MTT and transwell assays were applied to measure cell proliferation and motility. The survival rate was analyzed by Kaplan-Meier method accompanied with log-rank test. Dual luciferase assay was used to verify the relationship among miR-384 and CDK6 in ccRCC cells. MiR-384 was downregulated in ccRCC tissues and cell lines, and its overexpression inhibited cell proliferation and motility in ccRCC. In addition, miR-384 was found to be a marker for good prognosis in ccRCC. Furthermore, CDK6 was confirmed to be directly targeted by miR-384, and upregulation of CDK6 restored the inhibitory effects of miR- 384 in ccRCC. MiR-384 repressed tumorigenesis by regulating CDK6 in ccRCC and predicted good prognosis for ccRCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK6
Pricing and availability is not currently available.
Sigma-Aldrich
(Methylcyclopentadienyl)manganese(I) tricarbonyl
Pricing and availability is not currently available.