MilliporeSigma
  • Trastuzumab-Resistant HER2+ Breast Cancer Cells Retain Sensitivity to Poly (ADP-Ribose) Polymerase (PARP) Inhibition.
Trastuzumab-Resistant HER2+ Breast Cancer Cells Retain Sensitivity to Poly (ADP-Ribose) Polymerase (PARP) Inhibition.
Molecular cancer therapeutics (2018-03-30)
Monica E Wielgos, Zhuo Zhang, Rajani Rajbhandari, Tiffiny S Cooper, Ling Zeng, Andres Forero, Francisco J Esteva, C Kent Osborne, Rachel Schiff, Albert F LoBuglio, Susan E Nozell, Eddy S Yang
ABSTRACT
HER2-targeted therapies, such as trastuzumab, have increased the survival rates of HER2+ breast cancer patients. However, despite these therapies, many tumors eventually develop resistance to these therapies. Our lab previously reported an unexpected sensitivity of HER2+ breast cancer cells to poly (ADP-ribose) polymerase inhibitors (PARPi), agents that target homologous recombination (HR)-deficient tumors, independent of a DNA repair deficiency. In this study, we investigated whether HER2+ trastuzumab-resistant (TR) breast cancer cells were susceptible to PARPi and the mechanism behind PARPi induced cytotoxicity. We demonstrate that the PARPi ABT-888 (veliparib) decreased cell survival in vitro and tumor growth in vivo of HER2+ TR breast cancer cells. PARP-1 siRNA confirmed that cytotoxicity was due, in part, to PARP-1 inhibition. Furthermore, PARP-1 silencing had variable effects on the expression of several NF-κB-regulated genes. In particular, silencing PARP-1 inhibited NF-κB activity and reduced p65 binding at the IL8 promoter, which resulted in a decrease in IL8 mRNA and protein expression. Our results provide insight in the potential mechanism by which PARPi induces cytotoxicity in HER2+ breast cancer cells and support the testing of PARPi in patients with HER2+ breast cancer resistant to trastuzumab. Mol Cancer Ther; 17(5); 921-30. ©2018 AACR.
Sigma-Aldrich
MISSION® esiRNA, targeting human PARP1