MilliporeSigma
  • 18F-Flortaucipir Binding in Choroid Plexus: Related to Race and Hippocampus Signal.
18F-Flortaucipir Binding in Choroid Plexus: Related to Race and Hippocampus Signal.
Journal of Alzheimer's disease : JAD (2018-04-05)
Christopher M Lee, Heidi I L Jacobs, Marta Marquié, John A Becker, Nicolas V Andrea, David S Jin, Aaron P Schultz, Matthew P Frosch, Teresa Gómez-Isla, Reisa A Sperling, Keith A Johnson
ABSTRACT
On target 18F-Flortaucipir (FTP) binding of Alzheimer's disease tau aggregates and off-target binding of melanocytes have been demonstrated with autoradiography. We aimed to investigate the hypothesis that if binding in choroid plexus (CP) is due to melanocytes, the signal would be elevated in Black/African American (B/AA) compared to White (W) participants. In addition, we examined whether CP signal affects measurements in adjacent regions, and whether correcting for spill-in effects has an influence on associations between hippocampus (HC) FTP and amyloid or cognition. FTP race differences in 147 Harvard Aging Brain Study participants (23 B/AA, 124W) were examined in CP, HC, HC covaried for CP, amygdala, inferior temporal gyrus, entorhinal cortex, and fusiform regions. Associations between CP FTP and other regions-of-interest (ROIs) were probed to assess spill-in effects. A statistical regression approach to attenuate CP spill-in was tested by relating adjusted HC SUVR residuals and unadjusted HC SUVR to race, cognition and amyloid. All analyses were covaried for age, sex, education and amyloid deposition, and Bonferroni-corrected for multiple comparisons. B/AA individuals had elevated CP and HC SUVR (p < 0.007), whereas other ROI SUVR and HC SUVR covaried for CP SUVR did not show race differences (p > 0.05). CP SUVR was associated with HC SUVR (p < 10-14), but with no other ROI SUVR (p > 0.05). When adjusting HC SUVR for CP SUVR, no race differences in residual HC SUVR were detected, and relationships with amyloid and memory became apparent. Melanocyte FTP binding may account partially for high CP signal. This off-target binding affects mainly HC FTP measurements, which should be interpreted with caution.
Sigma-Aldrich
PiB, ≥98% (HPLC)