MilliporeSigma
  • Flavonoids and triterpenes from Combretum fragrans with anti-inflammatory, antioxidant and antidiabetic potential.
Flavonoids and triterpenes from Combretum fragrans with anti-inflammatory, antioxidant and antidiabetic potential.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2018-02-17)
Amadou Dawé, Marius Mbiantcha, Fawai Yakai, Almas Jabeen, Muhammad Shaiq Ali, Mehreen Lateef, Bonaventure Tchaleu Ngadjui
ABSTRACT
Despite the well-documented benefits of Combretum fragrans in Cameroon, only few scientific works have been done on it. In this study we isolated eight compounds from the leaves extract of C. fragrans: velutin (1), belamcanidin (2), cirsilineol (3), cirsimaritin (4), 3β-acetoxy-20,24-epoxy-11,25-hydroxy-dammarane (5), combretin A (6), combretin B (7) and a mixture of arjunolic acid (8a) and asiatic acid (8b). Compounds 6 and 7 presented potent anti-inflammatory, antioxidant and antidiabetic activities. Compounds 1, 3, 5 and the mixture of 8a and 8b were significantly active, and compounds 2 and 4 presented moderate activity for reactive oxygen species inhibitory and free-radical scavenging. All compounds were isolated using chromatographic techniques; their structures were elucidated by spectroscopic techniques and their spectroscopic data compared with those of the literature. Anti-inflammatory activity was evaluated via the oxidative burst assay using a luminol-amplified chemiluminescence technique, antioxidant activity by free-radical scavenging activity (DPPH) and antidiabetic activity via α-glucosidase inhibition. All of the isolated compounds (1-8) were reported to exhibit significant antioxidant activity. Compounds 1, 3, and 5-8 exhibited potent chemiluminescence inhibition effect, and only compounds 6 and 7 inhibited α-glucosidase. Thus, C. fragrans can be used as an effective natural source of anti-inflammatory, antioxidant and antidiabetic compounds.
Sigma-Aldrich
Cerium(IV) sulfate