MilliporeSigma
  • Visiting the Limits between a Highly Strained 1-Zirconacyclobuta-2,3-diene and Chemically Robust Dizirconacyclooctatetraene.
Visiting the Limits between a Highly Strained 1-Zirconacyclobuta-2,3-diene and Chemically Robust Dizirconacyclooctatetraene.
Chemistry (Weinheim an der Bergstrasse, Germany) (2018-02-02)
Fabian Reiß, Melanie Reiß, Anke Spannenberg, Haijun Jiao, Wolfgang Baumann, Perdita Arndt, Uwe Rosenthal, Torsten Beweries
ABSTRACT
The reaction of the allene precursor Li2 (Me3 SiC3 SiMe3 ) with [Cp2 ZrCl2 ] (Cp=cyclopentadienyl) was examined. The selective formation of hitherto unknown linear, allene-bridged dizirconocene complexes [(Cp2 ZrCl)2 {-μ-(Me3 Si)C3 (SiMe3 )-}] and [(Cp2 Zr)2 {-μ-(Me3 Si)C3 (SiMe3 )-}2 ] was observed. Upon σ coordination of the allenediyl unit to {Cp2 Zr}, pyrophoric Li2 (Me3 SiC3 SiMe3 ) is tamed stepwise to yield a surprisingly robust 1,5-dizirconacyclooctatetra-2,3,6,7-ene with cumulated double bonds. This complex is unexpectedly inert against moisture, air, water and acetone. Surprisingly, it degrades under MS conditions to give the highly strained 1-zirconacyclobuta-2,3-diene. All compounds isolated have been fully characterised and the molecular structures are discussed. The stability and reactivity of these complexes are rationalised by DFT computations.
Sigma-Aldrich
Ethynyltrimethylsilane, 98%
Pricing and availability is not currently available.