MilliporeSigma
The direct anti-Markovnikov addition of mineral acids to styrenes.
Nature chemistry (2014-07-24)
Dale J Wilger, Jean-Marc M Grandjean, Taylor R Lammert, David A Nicewicz
ABSTRACT
The direct anti-Markovnikov addition of strong Brønsted acids to alkenes remains an unsolved problem in synthetic chemistry. Here, we report an efficient organic photoredox catalyst system for the addition of HCl, HF and also phosphoric and sulfonic acids to alkenes, with complete regioselectivity. These transformations were developed using a photoredox catalyst in conjunction with a redox-active hydrogen atom donor. The nucleophile counterion plays a critical role by ensuring high reactivity, with 2,6-lutidinium salts typically furnishing the best results. The nature of the redox-active hydrogen atom donor is also consequential, with 4-methoxythiophenol providing the best reactivity when 2,6-lutidinium salts are used. A novel acridinium sensitizer provides enhanced reactivity within several of the more challenging reaction manifolds. This Article demonstrates how nucleophilic addition reactions mediated by photoredox catalysis can change the way electrophilic and homofugal precursors are constructed.
Sigma-Aldrich
2,7-Dimethyl-9-mesityl-10-methylacridinium tetrafluoroborate
Pricing and availability is not currently available.