MilliporeSigma
  • A comparative study of the rheological and sensory properties of a petroleum-free and a petroleum-based cosmetic cream.
A comparative study of the rheological and sensory properties of a petroleum-free and a petroleum-based cosmetic cream.
Journal of cosmetic science (2018-04-06)
Fan C Wang, Alejandro G Marangoni
ABSTRACT
A petroleum-free skin cream was developed using food-grade ingredients. The rheological and sensorial properties of this petroleum-free skin cream were compared to a commercially available petroleum-based skin cream. Specifically, large-amplitude oscillatory shear (LAOS) characterization of the two skin creams was performed. The petroleum-free skin cream showed similar linear and nonlinear viscoelastic rheological properties, comparable skin hydration functions, and consumer acceptance as the commercially available skin cream. A schematic diagram aiming to correlate the physical and sensorial properties of skin cream was also proposed at the end of the work. Results of this work could provide the cosmetic industry necessary information for the development of alternatives for petroleum-based skin creams.
Sigma-Aldrich
(2-Hydroxyethyl)urea, 95%
Sigma-Aldrich
1-Stearoyl-rac-glycerol, ≥99%