MilliporeSigma
  • A fast and easy screening method for voice disorders among teacher students.
A fast and easy screening method for voice disorders among teacher students.
Logopedics, phoniatrics, vocology (2001-07-04)
S Simberg, E Sala, A Laine, A M Rönnemaa
ABSTRACT
A recent study concerning voice disorders among future teachers showed that about 20% of the students had a voice disorder. Most of the disorders were organic. In the current study, we describe a voice-screening method that can be administered by the health care personnel in order to select students for further examination by the speech language therapist and/or phoniatrician. The screening method consists of a perceptual assessment of voice quality and a questionnaire concerning vocal symptoms. As criterion for further medical investigation and voice therapy, we selected a score of 35 mm or above on a visual analogue scale assessing Grade, i.e. overall grade of hoarseness and/or two or more weekly or more often occurring vocal symptoms. The results showed that health care personnel with some training in assessing voices using the questionnaire are competent to perform a rough voice screening on students.
Product Description
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.80 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 105 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 1.00 μm