Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Zinc
Synonym(s):
Zinc
Empirical Formula (Hill Notation):
Zn
CAS Number:
7440-66-6
Molecular Weight:
65.39
Products
Product Category
metals (1)
Brand
Supelco (5)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.08789
Zinc,
powder for analysis particle size < 45 µm EMSURE
®
Expand
View Pricing
1.08780
Zinc,
granular for analysis, particle size about 3-8 mm EMSURE
®
ISO
Expand
View Pricing
1.08774
Zinc,
dust particle size < 63 µm EMPLURA
®
Expand
View Pricing
1.08756
Zinc,
coarse powder for analysis suitable for filling of reductors, particle size about 0.3- 1.5 mm (14-50 mesh ASTM) EMSURE
®
Reag. Ph Eur
Expand
View Pricing
1.08782
Zinc,
sticks
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More