Skip to Content
MilliporeSigma
Valeryl Salicylate ≥98% (HPLC)

Valeryl Salicylate

Valeryl Salicylate ≥98% (HPLC)
Synonym(s):
2-Valeryloxybenzoic acid, Valerylsalicylic acid, vsa
Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24

Products

Shipping
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing