Skip to Content
MilliporeSigma
TSKgel® Alpha-3000 HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size

TSKgel® Alpha-3000 (7 µm) HPLC Columns

TSKgel® Alpha-3000 HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size
Synonym(s):
TSKgel® Alpha Columns

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing