Trypan blue (C.I. 23850)
Synonym(s):
Trypan Blue, Direct blue 14
Linear Formula:
C34H24N6O14S4Na4
CAS Number:
Molecular Weight:
960.81
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing