Skip to Content
MilliporeSigma
Tris(3,5-dimethylphenyl)phosphine 96%

Tris(3,5-dimethylphenyl)phosphine

Tris(3,5-dimethylphenyl)phosphine 96%
Synonym(s):
Tri-3,5-xylylphosphine
Linear Formula:
[(CH3)2C6H3]3P
CAS Number:
Molecular Weight:
346.44

Products

Shipping
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing