Skip to Content
MilliporeSigma
Tridodecylmethylammonium nitrate Selectophore™, ≥99.0%

Tridodecylmethylammonium nitrate

Tridodecylmethylammonium nitrate Selectophore™, ≥99.0%
Synonym(s):
Methyltridodecylammonium nitrate
Linear Formula:
[CH3(CH2)11]3N(NO3)CH3
CAS Number:
Molecular Weight:
599.03

Products

Shipping
Product Category
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing