Thiamine Disulfide
Synonym(s):
N-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-N-[(E)-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-hydroxypent-2-en-3-yl]disulfanyl]-5-hydroxypent-2-en-2-yl]formamide
Empirical Formula (Hill Notation):
C24H34N8O4S2
CAS Number:
Molecular Weight:
562.71
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing