Skip to Content
MilliporeSigma
Tetramethylrhodamine ethyl ester perchlorate suitable for fluorescence, ≥90% (HPCE)

Tetramethylrhodamine ethyl ester perchlorate

Tetramethylrhodamine ethyl ester perchlorate suitable for fluorescence, ≥90% (HPCE)
Synonym(s):
TMRE
Empirical Formula (Hill Notation):
C26H27ClN2O7
CAS Number:
Molecular Weight:
514.95

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing