
Sulfadimethoxine-d6

Empirical Formula (Hill Notation):
C12D6H8N4O4S
CAS Number:
Molecular Weight:
316.37
Products
Shipping
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1