Skip to Content
MilliporeSigma
Sodium acetate-13C2,d3 99 atom % D, 99 atom % 13C

Sodium acetate-13C2,d3

Sodium acetate-13C2,d3 99 atom % D, 99 atom % 13C
Synonym(s):
13C and D Labeled sodium acetate, Acetic-13C2,2,2,2-d3 acid sodium salt
Linear Formula:
13CD313CO2Na
CAS Number:
Molecular Weight:
87.04

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing