Skip to Content
MilliporeSigma
Sodium acetate-1-13C,d3 99 atom % 13C, 99 atom % D

Sodium acetate-1-13C,d3

Sodium acetate-1-13C,d3 99 atom % 13C, 99 atom % D
Synonym(s):
Acetic acid-1-13C-2,2,2-d3 sodium salt
Linear Formula:
CD313CO2Na
CAS Number:
Molecular Weight:
86.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing